Assay Details
Target Gene Details
Entrez Gene ID: | 18189 |
Gene Name: | neurexin I |
Gene Aliases: |
1700062G21Rik, 9330127H16Rik, A230068P09Rik, mKIAA0578 |
Location: |
Chr.17:90033644-91093100 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrxn1 | NM_020252.3 | 15 | 4035 | NP_064648.3 |
| NM_177284.2 | 13 | 3999 | NP_796258.2 | |
| AB093249.2 | 15 | 4035 | BAC41433.2 | |
| AK133271.1 | 10 | 2626 | ||
| AK137407.1 | 14 | 4019 |

Genomic Map