Assay Details
Target Gene Details
Entrez Gene ID: | 73884 |
Gene Name: | zinc finger, DBF-type containing 2 |
Gene Aliases: |
4930431J08Rik, 9330107J05Rik |
Location: |
Chr.1:63200358-63314576 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zdbf2 | NM_001285936.1 | NP_001272865.1 | ||
| NM_001285937.1 | NP_001272866.1 | |||
| AB777269.1 | ||||
| AB777270.1 |
More Information
Set Membership: |
|

Genomic Map