Assay Details
Target Gene Details
Entrez Gene ID: | 227290 |
Gene Name: | angio-associated migratory protein |
Gene Aliases: |
AU040907, Aamp-rs |
Location: |
Chr.1:74279840-74284738 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aamp | NM_001190444.1 | NP_001177373.1 | ||
| NM_146110.3 | NP_666222.2 | |||
| AK146470.1 | BAE27196.1 | |||
| AK151820.1 | BAE30717.1 | |||
| AK162750.1 | BAE37048.1 | |||
| AK167621.1 | BAE39673.1 | |||
| BC031795.1 | AAH31795.1 |

Genomic Map