Assay Details
Target Gene Details
Entrez Gene ID: | 266645 |
Gene Name: | amino carboxymuconate semialdehyde decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.1:127729413-127767564 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acmsd | NM_001033041.2 | NP_001028213.1 | ||
| AB071419.1 | BAB86939.1 | |||
| AK143962.1 | BAE25634.1 | |||
| BC139281.1 |
More Information
Set Membership: |
|

Genomic Map