Assay Details
Target Gene Details
Entrez Gene ID: | 100502745 |
Gene Name: | RIKEN cDNA 2900009J06 gene |
Gene Aliases: |
- |
Location: |
Chr.1:127753617-127774054 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2900009J06Rik | NR_045298.1 | |||
| AK013506.1 | ||||
| BB663189.1 | ||||
| BX636135.1 |
Target Gene Details
Entrez Gene ID: | 266645 |
Gene Name: | amino carboxymuconate semialdehyde decarboxylase |
Gene Aliases: |
- |
Location: |
Chr.1:127729413-127767564 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acmsd | NM_001033041.2 | 9 | 968 | NP_001028213.1 |
| AK143962.1 | 9 | 970 | BAE25634.1 | |
| AK165228.1 | 3 | 334 | BAE38088.1 | |
| BC139281.1 | 9 | 919 |
More Information
Set Membership: |
|

Genomic Map