Assay Details
Target Gene Details
Entrez Gene ID: | 73844 |
Gene Name: | ankyrin repeat domain 45 |
Gene Aliases: |
4933409K03Rik |
Location: |
Chr.1:161142547-161170507 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd45 | NM_028664.1 | 7 | 984 | NP_082940.1 |
| AK016757.1 | 6 | 903 | BAB30413.1 | |
| BC049713.1 | 7 | 956 | AAH49713.1 |
More Information
Set Membership: |
|

Genomic Map