Assay Details
Target Gene Details
Entrez Gene ID: | 211660 |
Gene Name: | centrosome and spindle pole associated protein 1 |
Gene Aliases: |
2310020J12Rik, 4930413O22Rik, AA387550, BC055915, Cspp2, Cspp3 |
Location: |
Chr.1:10038218-10136771 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cspp1 | NM_026493.3 | NP_080769.3 | ||
| BC151042.1 | ||||
| CN457021.1 | ||||
| FJ008676.1 | ||||
| FJ008677.1 |
More Information
Set Membership: |
|

Genomic Map