Assay Details
Target Gene Details
Entrez Gene ID: | 76707 |
Gene Name: | CLIP associating protein 1 |
Gene Aliases: |
1700030C23Rik, 5730583A19Rik, B130045P17Rik, CLASP1alpha |
Location: |
Chr.1:118389058-118609462 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clasp1 | NM_001293300.1 | 11 | 1728 | NP_001280229.1 |
| NM_001293301.1 | 11 | 1728 | NP_001280230.1 | |
| NM_029709.2 | 11 | 1728 | NP_083985.2 | |
| AK031181.1 | 11 | 1330 | ||
| AK080782.1 | 11 | 1444 | BAC38020.1 | |
| BC094432.1 | 10 | 1269 | ||
| BC158064.1 | 11 | 1331 |
More Information
Set Membership: |
|

Genomic Map