Assay Details
Target Gene Details
Entrez Gene ID: | 329260 |
Gene Name: | DENN/MADD domain containing 1B |
Gene Aliases: |
4632404N19Rik, 4930467M19Rik, 6820401H01Rik, F730008N07Rik |
Location: |
Chr.1:138963446-139176043 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 21 - Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd1b | NM_001166501.1 | NP_001159973.1 | ||
| NM_181347.3 | NP_851992.1 | |||
| AK015524.1 | BAB29881.1 | |||
| AK155727.1 | BAE33404.1 | |||
| BC141388.1 | ||||
| BC141389.1 |

Genomic Map