Assay Details
Target Gene Details
Entrez Gene ID: | 13838 |
Gene Name: | Eph receptor A4 |
Gene Aliases: |
2900005C20Rik, AI385584, Cek8, Hek8, Sek, Sek1, Tyro1, rb |
Location: |
Chr.1:77367182-77515103 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha4 | NM_007936.3 | NP_031962.2 | ||
| AK028509.1 | ||||
| AK147698.1 | BAE28081.1 | |||
| BC004782.1 | AAH04782.1 | |||
| BC052164.1 | AAH52164.1 | |||
| S57168.1 | AAB25836.1 | |||
| X65138.1 | CAA46268.1 |
More Information
Set Membership: |
|

Genomic Map