Assay Details
Target Gene Details
Entrez Gene ID: | 241112 |
Gene Name: | ciliogenesis associated TTC17 interacting protein |
Gene Aliases: |
Gm216 |
Location: |
Chr.1:74362108-74369323 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catip | NM_001033345.2 | NP_001028517.1 | ||
| AK138157.1 | BAE23563.1 |

Genomic Map