Assay Details
Target Gene Details
Entrez Gene ID: | 56489 |
Gene Name: | inhibitor of kappaB kinase epsilon |
Gene Aliases: |
AW558201, IKK-i, IKKepsilon, Ikki |
Location: |
Chr.1:131254600-131279625 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbke | NM_019777.3 | 2 | 275 | NP_062751.2 |
| AK088580.1 | 2 | 277 | BAC40434.1 | |
| AK149911.1 | 2 | 264 | BAE29161.1 | |
| AK153046.1 | 2 | 276 | BAE31675.1 |
More Information
Set Membership: |
|

Genomic Map