Assay Details
Target Gene Details
Entrez Gene ID: | 58181 |
Gene Name: | interleukin 20 |
Gene Aliases: |
Zcyto10 |
Location: |
Chr.1:130907192-130911296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il20 | NM_021380.1 | NP_067355.1 | ||
| AF224267.1 | AAF36680.1 | |||
| AK078698.1 | ||||
| AY876923.1 | AAW78349.1 | |||
| BC111821.1 | AAI11822.1 |
More Information
Set Membership: |
|

Genomic Map