Assay Details
Target Gene Details
Entrez Gene ID: | 93672 |
Gene Name: | interleukin 24 |
Gene Aliases: |
FISP, Mda-7, Mda7, St16 |
Location: |
Chr.1:130882074-130887413 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il24 | NM_053095.2 | NP_444325.2 | ||
| AF235006.1 | AAK52590.1 | |||
| AF333251.1 | AAK52470.1 | |||
| DQ401033.1 |

Genomic Map