Assay Details
Target Gene Details
Entrez Gene ID: | 227288 |
Gene Name: | chemokine (C-X-C motif) receptor 1 |
Gene Aliases: |
Il8ra |
Location: |
Chr.1:74191785-74198144 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcr1 | NM_178241.4 | NP_839972.1 | ||
| AY390263.1 | AAQ84305.1 |

Genomic Map