Assay Details
Target Gene Details
Entrez Gene ID: | 68338 |
Gene Name: | golgi transport 1 homolog A (S. cerevisiae) |
Gene Aliases: |
0610012C01Rik, Kiss1, metastatin |
Location: |
Chr.1:133309823-133323026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golt1a | NM_026680.4 | NP_080956.1 | ||
| AK002581.1 | BAB22205.1 | |||
| AK141378.1 | BAE24665.1 | |||
| AY707855.1 | AAW34128.1 | |||
| BC024448.1 | AAH24448.1 |
More Information
Set Membership: |
|

Genomic Map