Assay Details
Target Gene Details
Entrez Gene ID: | 246256 |
Gene Name: | Fc receptor, IgG, low affinity IV |
Gene Aliases: |
4833442P21Rik, CD16-2, FcgRIV, FcgammaRIV, Fcgr3a, Fcrl3 |
Location: |
Chr.1:171018926-171029761 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcgr4 | NM_144559.2 | NP_653142.2 | ||
| AK076532.1 | BAC36381.1 | |||
| AK087863.1 | ||||
| BC027310.1 | AAH27310.1 |
More Information
Set Membership: |
|

Genomic Map