Assay Details
Target Gene Details
Entrez Gene ID: | 70750 |
Gene Name: | 3-ketodihydrosphingosine reductase |
Gene Aliases: |
6330410P18Rik, 9430079B08Rik, Fvt1 |
Location: |
Chr.1:106720410-106759830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdsr | NM_027534.2 | NP_081810.1 | ||
| AK018155.1 | ||||
| AK170064.1 | BAE41541.1 | |||
| AY634684.1 | AAT57900.1 | |||
| BC023820.1 |
More Information
Set Membership: |
|

Genomic Map