Assay Details
Target Gene Details
Entrez Gene ID: | 76668 |
Gene Name: | malate dehydrogenase 1B, NAD (soluble) |
Gene Aliases: |
1700124B08Rik, AV255588 |
Location: |
Chr.1:63687573-63730547 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdh1b | NM_029696.4 | NP_083972.3 | ||
| AK007262.1 | BAB24922.1 | |||
| BC050786.1 | AAH50786.1 |
More Information
Set Membership: |
|

Genomic Map