Assay Details
Target Gene Details
Entrez Gene ID: | 70988 |
Gene Name: | RIKEN cDNA 4931428L18 gene |
Gene Aliases: |
4930563O14Rik |
Location: |
Chr.1:31141078-31222656 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931428L18Rik | NR_033445.1 | |||
| AK016483.1 | BAB30263.1 | |||
| BC147514.1 | ||||
| BC147546.1 | ||||
| BC172083.1 |
Target Gene Details
Entrez Gene ID: | 266744 |
Gene Name: | lengsin, lens protein with glutamine synthetase domain |
Gene Aliases: |
Gluld1, Lgs, lengsin |
Location: |
Chr.1:31176435-31204725 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgsn | NM_153601.1 | NP_705829.1 | ||
| AF545851.1 | AAN38298.1 | |||
| BC117745.1 | ||||
| BC117746.1 | ||||
| BC128067.1 |
More Information
Set Membership: |
|

Genomic Map