Assay Details
Target Gene Details
Entrez Gene ID: | 111175 |
Gene Name: | peroxisomal trans-2-enoyl-CoA reductase |
Gene Aliases: |
2400003B18Rik, TERP |
Location: |
Chr.1:72259172-72284314 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pecr | NM_023523.5 | NP_076012.3 | ||
| AF232011.1 | AAF69800.1 | |||
| AF242204.1 | AAK28336.1 | |||
| AK010260.1 | BAB26803.1 | |||
| AK090327.1 | ||||
| AK135714.1 | BAE22624.1 | |||
| AK162100.1 | ||||
| BC013530.1 | AAH13530.1 |
More Information
Set Membership: |
|

Genomic Map