Assay Details
Target Gene Details
Entrez Gene ID: | 67459 |
Gene Name: | nuclear VCP-like |
Gene Aliases: |
1200009I24Rik |
Location: |
Chr.1:181087140-181144215 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nvl | NM_026171.1 | NP_080447.1 | ||
| AK004676.1 | BAB23464.1 | |||
| AK054502.1 | BAC35806.1 | |||
| AK134620.1 | ||||
| AK140509.1 | BAE24409.1 | |||
| BC031847.1 | AAH31847.1 |

Genomic Map