Assay Details
Target Gene Details
Entrez Gene ID: | 329251 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 12B |
Gene Aliases: |
1810037O03Rik, 9530009M10Rik, AI132431, Mypt2 |
Location: |
Chr.1:134754665-134955965 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r12b | NM_001081307.1 | NP_001074776.1 | ||
| NR_015606.3 | ||||
| AK035234.1 | 1 | 1096 | ||
| AK035285.1 | ||||
| AK039445.1 | ||||
| AK051953.1 | ||||
| AK161046.1 | ||||
| BC145746.1 |
More Information
Set Membership: |
|

Genomic Map