Assay Details
Target Gene Details
Entrez Gene ID: | 329251 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 12B |
Gene Aliases: |
1810037O03Rik, 9530009M10Rik, AI132431, Mypt2 |
Location: |
Chr.1:134754665-134955965 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r12b | NM_001081307.1 | NP_001074776.1 | ||
| AK042847.1 | BAC31381.1 | |||
| AK046012.1 | BAC32572.1 | |||
| AK046167.1 | BAC32618.1 | |||
| AK080855.1 | BAC38046.1 | |||
| AK141375.1 | ||||
| BC145746.1 |
More Information
Set Membership: |
|

Genomic Map