Assay Details
Target Gene Details
Entrez Gene ID: | 19243 |
Gene Name: | protein tyrosine phosphatase 4a1 |
Gene Aliases: |
AA415290, AU019864, C130021B01, Prl-1 |
Location: |
Chr.1:30940303-30949765 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptp4a1 | NM_011200.2 | NP_035330.1 | ||
| AK081491.1 | BAC38233.1 | |||
| AK148288.1 | BAE28460.1 | |||
| AK150506.1 | BAE29619.1 | |||
| AK150556.1 | ||||
| AK150762.1 | ||||
| AK151003.1 | ||||
| AK151533.1 | BAE30480.1 | |||
| BC055039.1 | AAH55039.1 | |||
| BC086787.1 | AAH86787.1 | |||
| BC094447.1 | AAH94447.1 | |||
| BC150972.1 | ||||
| BC150981.1 | ||||
| U84411.1 | AAB58913.1 |
More Information
Set Membership: |
|

Genomic Map