Assay Details
Target Gene Details
Entrez Gene ID: | 19250 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 14 |
Gene Aliases: |
C130080N23Rik, OTTMUSG00000022087, PTP36 |
Location: |
Chr.1:189728216-189876695 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn14 | NM_008976.2 | NP_033002.2 | ||
| AF170902.1 | AAF27548.1 | |||
| AF170903.1 | AAF27549.1 | |||
| AF170904.1 | AAF27550.1 | |||
| AF170905.1 | AAF27551.1 | |||
| AK028783.1 | BAC26118.1 | |||
| AK037646.1 | BAC29840.1 | |||
| AK086524.1 | BAC39685.1 | |||
| AK156480.1 | ||||
| D31842.1 | BAA06628.1 |
More Information
Set Membership: |
|

Genomic Map