Assay Details
Target Gene Details
Entrez Gene ID: | 20652 |
Gene Name: | sterol O-acyltransferase 1 |
Gene Aliases: |
8430426K15Rik, ACAT-1, AW550831, Acact, ald, hid |
Location: |
Chr.1:156428108-156474328 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Soat1 | NM_009230.3 | 12 | 1349 | NP_033256.2 |
| AK052299.1 | 9 | 906 | BAC34925.1 | |
| AK142486.1 | 12 | 1266 | BAE25081.1 | |
| AK159393.1 | 12 | 1279 | BAE35047.1 | |
| AK159529.1 | 12 | 1279 | BAE35158.1 | |
| AK160017.1 | 12 | 1279 | BAE35563.1 | |
| BC083092.1 | 11 | 1163 | AAH83092.1 | |
| DQ903181.1 | 12 | 1266 | ||
| S81092.1 | 12 | 1283 | AAB36050.1 |
More Information
Set Membership: |
|

Genomic Map