Assay Details
Target Gene Details
Entrez Gene ID: | 69453 |
Gene Name: | protease, serine 56 |
Gene Aliases: |
1700027L20Rik |
Location: |
Chr.1:87183094-87188405 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss56 | NM_027084.1 | NP_081360.1 | ||
| JF323950.1 | ||||
| JF698684.1 |
More Information
Set Membership: |
|

Genomic Map