Assay Details
Target Gene Details
Entrez Gene ID: | 11363 |
Gene Name: | acyl-Coenzyme A dehydrogenase, long-chain |
Gene Aliases: |
AA960361, AU018452, C79855, LCAD |
Location: |
Chr.1:66830839-66863309 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acadl | NM_007381.4 | 6 | 751 | NP_031407.2 |
| AK005140.1 | 6 | 710 | BAB23838.1 | |
| AK018319.1 | 5 | 527 | BAB31161.1 | |
| AK151845.1 | 6 | 720 | ||
| AK167537.1 | 6 | 718 | ||
| AK187227.1 | ||||
| AK216184.1 | 3 | 204 | ||
| BC027412.1 | 6 | 708 | AAH27412.1 | |
| U21489.1 | 6 | 691 | AAC52329.1 |
More Information
Set Membership: |
|

Genomic Map