Assay Details
Target Gene Details
Entrez Gene ID: | 70988 |
Gene Name: | RIKEN cDNA 4931428L18 gene |
Gene Aliases: |
4930563O14Rik |
Location: |
Chr.1:31141078-31222656 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931428L18Rik | NR_033445.1 | 1 | 120 | |
| AK016212.1 | 1 | 121 | BAB30149.1 | |
| AK016483.1 | 1 | 76 | BAB30263.1 | |
| AK162175.1 | 1 | 77 | ||
| BC147514.1 | 1 | 92 | ||
| BC147546.1 | 1 | 92 | ||
| BC172083.1 | 1 | 92 |

Genomic Map