Assay Details
Target Gene Details
Entrez Gene ID: | 619326 |
Gene Name: | RIKEN cDNA 9130409I23 gene |
Gene Aliases: |
- |
Location: |
Chr.1:181006612-181060761 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9130409I23Rik | NM_001033819.2 | NP_001028991.1 | ||
| AK136575.1 | BAE23057.1 | |||
| AK162201.1 | BAE36788.1 | |||
| BC147378.1 | ||||
| BC147379.1 |
More Information
Set Membership: |
|

Genomic Map