Assay Details
Target Gene Details
Entrez Gene ID: | 227290 |
Gene Name: | angio-associated migratory protein |
Gene Aliases: |
AU040907, Aamp-rs |
Location: |
Chr.1:74279840-74284738 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aamp | NM_001190444.1 | 2 | 312 | NP_001177373.1 |
| NM_146110.3 | 2 | 309 | NP_666222.2 | |
| AK146470.1 | 2 | 252 | BAE27196.1 | |
| AK151820.1 | 2 | 221 | BAE30717.1 | |
| AK162750.1 | 2 | 237 | BAE37048.1 | |
| AK167621.1 | 2 | 233 | BAE39673.1 | |
| AK194894.1 | ||||
| AK199042.1 | ||||
| AK200082.1 | ||||
| AK215002.1 | ||||
| BC031795.1 | AAH31795.1 | |||
| BY101182.1 | 2 | 309 |

Genomic Map