Assay Details
Target Gene Details
Entrez Gene ID: | 381290 |
Gene Name: | ATPase, Ca++ transporting, plasma membrane 4 |
Gene Aliases: |
Pmca4 |
Location: |
Chr.1:133702674-133801037 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp2b4 | NM_001167949.2 | 19 | 3691 | NP_001161421.1 |
| NM_213616.4 | 19 | 3691 | NP_998781.1 | |
| AK138298.1 | ||||
| AY560895.1 | 19 | 3225 | AAT01506.1 | |
| AY560896.1 | AAT01507.1 | |||
| BC109172.1 | 19 | 3225 | AAI09173.1 | |
| BC109173.1 | 19 | 3225 | AAI09174.1 | |
| EU493094.2 | 19 | 3395 | ||
| EU493095.2 | 19 | 3395 | ||
| JF683384.1 | 19 | 3395 |
More Information
Set Membership: |
|

Genomic Map