Assay Details
Target Gene Details
Entrez Gene ID: | 381290 |
Gene Name: | ATPase, Ca++ transporting, plasma membrane 4 |
Gene Aliases: |
Pmca4 |
Location: |
Chr.1:133702674-133801037 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp2b4 | NM_001167949.2 | NP_001161421.1 | ||
| NM_213616.4 | NP_998781.1 | |||
| AK143672.1 | BAE25488.1 | |||
| AY560895.1 | AAT01506.1 | |||
| BC109172.1 | AAI09173.1 | |||
| BC109173.1 | AAI09174.1 | |||
| EU493094.2 | ||||
| EU493095.2 | ||||
| JF683384.1 |
More Information
Set Membership: |
|

Genomic Map