Assay Details
Target Gene Details
Entrez Gene ID: | 211673 |
Gene Name: | ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldin A-inhibited) |
Gene Aliases: |
ARFGEP1, BIG1, D130059B05Rik, D730028O18Rik, P200 |
Location: |
Chr.1:10137507-10232670 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgef1 | NM_001102430.1 | 36 | 5372 | NP_001095900.1 |
| AK035885.1 | 4 | 414 | ||
| AK051592.1 | 7 | 878 | BAC34685.1 | |
| BC025221.1 | 4 | 514 | AAH25221.1 |
More Information
Set Membership: |
|

Genomic Map