Assay Details
Target Gene Details
Entrez Gene ID: | 12334 |
Gene Name: | calpain 2 |
Gene Aliases: |
AI326419, Capa-2, Capa2, m-calpain, m-calpin |
Location: |
Chr.1:182467259-182517483 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capn2 | NM_009794.3 | 10 | 1393 | NP_033924.2 |
| AF015038.1 | 10 | 1339 | AAB94029.1 | |
| AK052141.1 | BAC34855.1 | |||
| AK053832.1 | 10 | 2679 | ||
| AK078787.1 | 6 | 2022 | ||
| AK150695.1 | 10 | 1367 | BAE29774.1 | |
| AK152094.1 | 10 | 1367 | BAE30942.1 | |
| AK152790.1 | 10 | 1370 | ||
| AK152994.1 | 10 | 1370 | ||
| AK154855.1 | 10 | 1407 | BAE32880.1 | |
| AK181353.1 | ||||
| AK207878.1 | ||||
| BC054726.1 | 10 | 1393 | AAH54726.1 | |
| D38117.1 | 10 | 1434 | BAA22964.1 | |
| Y10139.1 | 10 | 1275 | CAA71227.1 |
More Information
Set Membership: |
|

Genomic Map