Assay Details
Target Gene Details
Entrez Gene ID: | 67426 |
Gene Name: | aarF domain containing kinase 3 |
Gene Aliases: |
4632432J16Rik, AI462003, Cabc1, mKIAA0451 |
Location: |
Chr.1:180165238-180196020 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adck3 | NM_001163290.1 | 12 | 1499 | NP_001156762.1 |
| NM_023341.3 | 12 | 1597 | NP_075830.2 | |
| AJ278735.1 | 12 | 1477 | CAC00538.1 | |
| AK004177.1 | 5 | 482 | ||
| AK004791.1 | 12 | 1539 | BAB23567.2 | |
| AK014605.2 | 12 | 1539 | BAB29459.2 | |
| AK186293.1 | 3 | 223 | ||
| AK191635.1 | 3 | 222 | ||
| AK192575.1 | 3 | 223 | ||
| AK194890.1 | 3 | 222 | ||
| AK196005.1 | 3 | 223 | ||
| AK200253.1 | 3 | 222 | ||
| AK202430.1 | 3 | 222 | ||
| AK205042.1 | ||||
| AK207786.1 | 3 | 222 | ||
| AK217974.1 | 4 | 337 | ||
| BC030937.1 | 12 | 1545 | AAH30937.1 | |
| U31629.1 | 10 | 1138 | AAA86413.1 |

Genomic Map