Assay Details
Target Gene Details
Entrez Gene ID: | 12654 |
Gene Name: | chitinase-like 1 |
Gene Aliases: |
AW208766, Brp39, Chi3l1, Gp39 |
Location: |
Chr.1:134182164-134190031 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chil1 | NM_007695.3 | NP_031721.2 | ||
| CJ147225.1 |
More Information
Set Membership: |
|

Genomic Map