Assay Details
Target Gene Details
Entrez Gene ID: | 12654 |
Gene Name: | chitinase-like 1 |
Gene Aliases: |
AW208766, Brp39, Chi3l1, Gp39 |
Location: |
Chr.1:134182164-134190031 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chil1 | NM_007695.3 | NP_031721.2 | ||
| AK051475.1 | BAC34654.1 | |||
| AK155412.1 | BAE33251.1 | |||
| BC003780.1 | AAH03780.1 | |||
| BC004734.1 | AAH04734.1 | |||
| BC005611.1 | AAH05611.1 | |||
| CT010283.1 | CAJ18491.1 | |||
| EU313768.1 | ||||
| X93035.1 | CAA63603.1 |
More Information
Set Membership: |
|

Genomic Map