Assay Details
Target Gene Details
Entrez Gene ID: | 76707 |
Gene Name: | CLIP associating protein 1 |
Gene Aliases: |
1700030C23Rik, 5730583A19Rik, B130045P17Rik, CLASP1alpha |
Location: |
Chr.1:118389058-118609462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clasp1 | NM_001293300.1 | NP_001280229.1 | ||
| NM_001293301.1 | NP_001280230.1 | |||
| NM_029709.2 | NP_083985.2 | |||
| AJ276962.1 | CAC35162.1 | |||
| AK031181.1 | ||||
| AK042402.1 | ||||
| AK080782.1 | BAC38020.1 | |||
| BC094432.1 | ||||
| BC158064.1 |
More Information
Set Membership: |
|

Genomic Map