Assay Details
Target Gene Details
Entrez Gene ID: | 103967 |
Gene Name: | dynamin 3 |
Gene Aliases: |
9630020E24Rik, AW061159, B230343F03Rik, C530045C17, mKIAA0820 |
Location: |
Chr.1:161982450-162478323 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnm3 | NM_001038619.1 | 18 | 2181 | NP_001033708.1 |
| NM_172646.2 | 17 | 2173 | NP_766234.1 | |
| AK036199.1 | 18 | 2146 | BAC29343.1 | |
| AK049724.1 | 17 | 2174 | BAC33895.1 | |
| AK082698.1 | 18 | 2125 | BAC38575.1 | |
| AK122379.1 | 9 | 793 | BAC65661.1 | |
| AK147678.1 | 18 | 2125 | BAE28068.1 | |
| AK164444.1 | 4 | 384 | ||
| BC141143.1 | 18 | 2029 | ||
| BC141144.1 | 18 | 2029 |
More Information
Set Membership: |
|

Genomic Map