Assay Details
Target Gene Details
Entrez Gene ID: | 320054 |
Gene Name: | RIKEN cDNA 9230116N13 gene |
Gene Aliases: |
- |
Location: |
Chr.1:136411916-136415519 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9230116N13Rik | NR_024328.1 | 1 | 231 | |
| AK033832.1 | 1 | 232 | ||
| AK076438.1 | 1 | 208 |
Target Gene Details
Entrez Gene ID: | 67997 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 59 |
Gene Aliases: |
1210002B07Rik, 4833411G06Rik |
Location: |
Chr.1:136415271-136456190 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx59 | NM_026500.3 | NP_080776.1 | ||
| AK004833.1 | BAB23601.1 | |||
| AK013179.1 | BAB28694.1 | |||
| AK076462.1 | BAC36354.1 | |||
| AK133731.1 | BAE21808.1 |

Genomic Map