Assay Details
Target Gene Details
Entrez Gene ID: | 227325 |
Gene Name: | delta/notch-like EGF-related receptor |
Gene Aliases: |
A930026D19Rik, BET, Bret |
Location: |
Chr.1:84369839-84696221 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dner | NM_152915.1 | NP_690879.1 | ||
| AB067650.1 | BAB72175.1 | |||
| AF370126.1 | AAM14419.1 | |||
| AK044597.1 | BAC31995.1 | |||
| AK152755.1 | BAE31470.1 | |||
| AK153235.1 | BAE31828.1 | |||
| AY032924.1 | AAK50342.1 | |||
| BC022636.1 | AAH22636.1 | |||
| BC034634.1 | AAH34634.1 |

Genomic Map