Assay Details
Target Gene Details
Entrez Gene ID: | 26381 |
Gene Name: | estrogen-related receptor gamma |
Gene Aliases: |
ERR3, Errg, NR3B3, mKIAA0832 |
Location: |
Chr.1:187608792-188214884 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrrg | NM_001243792.1 | 6 | 1052 | NP_001230721.1 |
| NM_011935.3 | 5 | 990 | NP_036065.1 | |
| AB093266.1 | 6 | 1071 | BAC41450.1 | |
| AB221580.1 | 5 | 824 | ||
| AF117254.1 | 5 | 887 | AAD48369.1 | |
| AK052731.1 | 6 | 1031 | BAC35120.1 | |
| AK140676.1 | 3 | 339 | BAE24441.1 | |
| BC082324.1 | 5 | 958 | AAH82324.1 |

Genomic Map