Assay Details
Target Gene Details
Entrez Gene ID: | 13849 |
Gene Name: | epoxide hydrolase 1, microsomal |
Gene Aliases: |
AI195553, Eph-1, Eph1, Epxh1, mEH |
Location: |
Chr.1:180989556-181017495 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephx1 | NM_010145.2 | 2 | 129 | NP_034275.1 |
| AK018249.1 | 2 | 138 | BAB31132.1 | |
| BC029636.1 | 2 | 128 | AAH29636.1 | |
| BC045194.1 | 2 | 129 | AAH45194.1 | |
| BC057857.1 | 2 | 124 | AAH57857.1 | |
| BC061493.1 | 2 | 155 | AAH61493.1 | |
| U89491.1 | 2 | 98 | AAB49762.1 |
More Information
Set Membership: |
|

Genomic Map