Assay Details
Target Gene Details
Entrez Gene ID: | 13849 |
Gene Name: | epoxide hydrolase 1, microsomal |
Gene Aliases: |
AI195553, Eph-1, Eph1, Epxh1, mEH |
Location: |
Chr.1:180989556-181017495 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephx1 | NM_010145.2 | NP_034275.1 | ||
| AK018249.1 | BAB31132.1 | |||
| AK177895.1 | ||||
| AK180321.1 | ||||
| AK183956.1 | ||||
| AK204221.1 | ||||
| AK206265.1 | ||||
| AK207217.1 | ||||
| AK207438.1 | ||||
| AK207528.1 | ||||
| AK207577.1 | ||||
| AK207857.1 | ||||
| AK208317.1 | ||||
| AK208856.1 | ||||
| AK208963.1 | ||||
| AK209452.1 | ||||
| AK209650.1 | ||||
| AK210728.1 | ||||
| AK211418.1 | ||||
| AK211501.1 | ||||
| AK211877.1 | ||||
| AK211958.1 | ||||
| AK213096.1 | ||||
| AK214384.1 | ||||
| AK216019.1 | ||||
| AK216442.1 | ||||
| AK217465.1 | ||||
| AK218715.1 | ||||
| AK219272.1 | ||||
| AK219593.1 | ||||
| BC029636.1 | AAH29636.1 | |||
| BC045194.1 | AAH45194.1 | |||
| BC057857.1 | AAH57857.1 | |||
| BC061493.1 | AAH61493.1 | |||
| U89491.1 | AAB49762.1 |
More Information
Set Membership: |
|

Genomic Map