Assay Details
Target Gene Details
Entrez Gene ID: | 80915 |
Gene Name: | dual specificity phosphatase 12 |
Gene Aliases: |
1190004O14Rik, AA027408, AW049275, ESTM36, LMW-DSP4, T-DSP4, VH1, mVH1 |
Location: |
Chr.1:170874187-170885711 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp12 | NM_023173.2 | 2 | 534 | NP_075662.2 |
| AF268196.1 | 2 | 459 | AAG44739.1 | |
| AF280810.1 | 2 | 401 | AAK69508.1 | |
| AF280811.1 | 2 | 401 | AAK69509.1 | |
| AK004488.1 | 2 | 416 | BAB23328.1 | |
| AK155026.1 | 2 | 1533 | ||
| AK181232.1 | 2 | 192 | ||
| BC099453.1 | 2 | 534 | AAH99453.1 |
More Information
Set Membership: |
|

Genomic Map