Assay Details
Target Gene Details
Entrez Gene ID: | 69169 |
Gene Name: | Fc fragment of IgM receptor |
Gene Aliases: |
1810037B05Rik, Faim3, FcmuR, Toso |
Location: |
Chr.1:130865777-130880790 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcmr | NM_026976.2 | NP_081252.1 | ||
| AK007714.1 | BAB25207.1 | |||
| AK155943.1 | BAE33514.1 | |||
| AY227385.1 | ||||
| AY227386.1 | ||||
| AY227387.1 | ||||
| AY227388.1 | ||||
| AY227389.1 | ||||
| AY227390.1 | ||||
| AY227391.1 | ||||
| AY227392.1 | ||||
| AY227393.1 | ||||
| BC116674.1 | ||||
| BC118505.1 |
More Information
Set Membership: |
|

Genomic Map