Assay Details
Target Gene Details
Entrez Gene ID: | 226356 |
Gene Name: | cilia and flagella associated protein 221 |
Gene Aliases: |
Gm101, Pcdp1 |
Location: |
Chr.1:119923340-119997471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfap221 | NM_001115074.1 | NP_001108546.1 | ||
| EF632061.1 |
More Information
Set Membership: |
|

Genomic Map