Assay Details
Target Gene Details
Entrez Gene ID: | 23874 |
Gene Name: | phenylalanyl-tRNA synthetase, beta subunit |
Gene Aliases: |
C76708, Farsa, Farsl, Farslb, Frsb |
Location: |
Chr.1:78417958-78488897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Farsb | NM_001278075.1 | NP_001265004.1 | ||
| NM_011811.4 | NP_035941.2 | |||
| NR_102757.1 | ||||
| NR_102758.1 | ||||
| AF123263.1 | AAD26855.1 | |||
| AK010271.1 | BAB26810.1 | |||
| AK012154.1 | BAB28064.1 | |||
| AK045432.1 | BAC32363.1 | |||
| AK076607.1 | BAC36412.1 | |||
| AK134799.1 | BAE22290.1 | |||
| AK165645.1 | BAE38313.1 | |||
| AK167945.1 | BAE39948.1 | |||
| AK168923.1 | BAE40736.1 | |||
| AK169621.1 | ||||
| BC002313.1 | ||||
| BC016428.1 | AAH16428.1 | |||
| CN693684.1 |
More Information
Set Membership: |
|

Genomic Map